siRNA Details
Gene Symbol: BUB1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001211.5 | NP_001202.4 | 22, 23 | 3178 | ||
GenBank mRNA | AB208782.1 | - | 22, 23 | 3091 | ||
AF035933.1 | - | 22, 23 | 3087 | |||
AF046079.2 | - | 22, 23 | 3008 | |||
AF046918.1 | - | 22, 23 | 3096 | |||
AF053306.1 | - | 22, 23 | 3150 | |||
AF068760.1 | - | 22, 23 | 3072 | |||
AF107297.1 | - | 22, 23 | 3103 | |||
AK296795.1 | - | 22, 23 | 2793 | |||
AK296984.1 | - | 22, 23 | 3162 | |||
AK312709.1 | - | 22, 23 | 3146 | |||
BC018739.1 | - | 22, 23 | 3100 | 4649881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
