siRNA Details
Gene Symbol: BUB1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001211.5 | NP_001202.4 | 6 | 890 | ||
GenBank mRNA | AB208782.1 | - | 6 | 803 | ||
AF046079.2 | - | 6 | 720 | |||
AF046918.1 | - | 6 | 808 | |||
AF053306.1 | - | 6 | 862 | |||
AF068760.1 | - | 6 | 784 | |||
AF107297.1 | - | 6 | 815 | |||
AK296795.1 | - | 6 | 694 | |||
AK296984.1 | - | 6 | 874 | |||
AK312709.1 | - | 6 | 858 | |||
BC018739.1 | - | 6 | 812 | 4649881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
