siRNA Details
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258.2 | NP_001249.1 | 6 | 647 | ||
GenBank mRNA | X66357.1 | - | 5 | 659 |
Gene Symbol: TEN1-CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037709.1 | 8 | 2407 | |||
GenBank mRNA | AK097104.1 | - | 8 | 2364 | ||
BX647274.1 | - | 8 | 2393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
