siRNA Details
Gene Symbol: CDK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145306.1 | NP_001138778.1 | 8 | 2134 | ||
NM_001259.6 | NP_001250.1 | 8 | 2029 | |||
XM_006715835.2 | XP_006715898.1 | 8 | 2254 | |||
GenBank mRNA | BC052264.1 | - | 8 | 2010 | 6377987 | |
BQ048881.1 | - | 407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
