siRNA Details
Gene Symbol: CDK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260.2 | NP_001251.1 | 12 | 1677 | ||
NM_001318368.1 | NP_001305297.1 | 12 | 1674 | |||
XM_005266216.4 | XP_005266273.1 | 12 | 1035 | |||
XM_011534864.1 | XP_011533166.1 | 11 | 1598 | |||
XM_011534865.2 | XP_011533167.1 | 8 | 1722 | |||
XM_017020317.1 | XP_016875806.1 | 11 | 1033 | |||
XM_017020318.1 | XP_016875807.1 | 8 | 764 | |||
GenBank mRNA | AK296084.1 | - | 8 | 667 | ||
AK296875.1 | - | 11 | 1017 | |||
AY702976.1 | - | 11 | 1081 | |||
BC069634.1 | - | 12 | 1159 | 7262421 | ||
BC094784.1 | - | 11 | 1619 | |||
BC104492.1 | - | 12 | 1165 | 40032854 | ||
BC105950.1 | - | 12 | 1162 | 40032852 | ||
BC107601.1 | - | 12 | 1161 | 40032849 | ||
L23208.1 | - | 4 | 275 | |||
X85753.1 | - | 12 | 1165 |
Gene Symbol: ADRA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309989.1 | - | 1398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
