siRNA Details
Gene Symbol: CHEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114121.2 | NP_001107593.1 | 9 | 1781 | ||
NM_001114122.2 | NP_001107594.1 | 9 | 1781 | |||
NM_001244846.1 | NP_001231775.1 | 9 | 1781 | |||
NM_001274.5 | NP_001265.2 | 9 | 1124 | |||
NR_045204.1 | 8 | 1557 | ||||
NR_045205.1 | 9 | 1318 | ||||
XM_011542560.2 | XP_011540862.1 | 8 | 1783 | |||
XM_011542561.2 | XP_011540863.1 | 8 | 1783 | |||
XM_011542562.2 | XP_011540864.1 | 8 | 1033 | |||
XM_011542563.2 | XP_011540865.1 | 8 | 1098 | |||
XM_017017146.1 | XP_016872635.1 | 9 | 968 | |||
GenBank mRNA | AB451222.1 | - | 8 | 884 | ||
AB451345.1 | - | 8 | 884 | |||
AF016582.1 | - | 8 | 918 | |||
AF032874.1 | - | 8 | 884 | |||
AK292549.1 | - | 9 | 1318 | |||
AK293143.1 | - | 9 | 946 | |||
AK299783.1 | - | 8 | 718 | |||
AK300082.1 | - | 8 | 967 | |||
BC004202.2 | - | 9 | 943 | 3530606 | ||
BC017575.1 | - | 9 | 1042 | 4091997 | ||
JF289264.1 | - | 8 | 759 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
