siRNA Details
Gene Symbol: DHX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001357.4 | NP_001348.2 | 27 | 3624 | ||
NR_033302.1 | 28 | 3761 | ||||
GenBank mRNA | AB451248.1 | - | 3 | 344 | ||
AB451372.1 | - | 3 | 344 | |||
AK096582.1 | - | 8 | 1854 | |||
AK130564.1 | - | 3 | 520 | |||
AK226102.1 | - | 28 | 3742 | |||
BC058896.1 | - | 6 | 847 | |||
BC107881.1 | - | 7 | 968 | |||
BC137136.1 | - | 27 | 3571 | 9021133 | ||
BC144494.1 | - | 26 | 3497 | |||
L13848.1 | - | 27 | 3526 | |||
U03643.1 | - | 6 | 703 | |||
Y10658.1 | - | 26 | 3449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
