siRNA Details
Gene Symbol: HDAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001527.3 | NP_001518.3 | 12 | 1586 | ||
NR_033441.1 | 13 | 1614 | ||||
NR_073443.1 | 12 | 1492 | ||||
XM_011535788.1 | XP_011534090.1 | 13 | 1338 | |||
XM_017010799.1 | XP_016866288.1 | 13 | 1560 | |||
GenBank mRNA | AB209190.1 | - | 6 | 4302 | ||
AK092156.1 | - | 12 | 1492 | |||
AK097376.1 | - | 13 | 1548 | |||
AK296856.1 | - | 12 | 1586 | |||
BC031055.1 | - | 12 | 1550 | |||
U31814.1 | - | 12 | 1503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
