siRNA Details
Gene Symbol: EZH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321079.1 | NP_001308008.1 | 5 | 485 | ||
NM_001321081.1 | NP_001308010.1 | 6 | 549 | |||
NM_001321082.1 | NP_001308011.1 | 5 | 456 | |||
NM_001991.4 | NP_001982.2 | 6 | 576 | |||
XM_005257145.2 | XP_005257202.1 | 4 | 331 | |||
XM_011524517.2 | XP_011522819.1 | 5 | 424 | |||
XM_017024350.1 | XP_016879839.1 | 5 | 403 | |||
XM_017024351.1 | XP_016879840.1 | 4 | 310 | |||
XM_017024352.1 | XP_016879841.1 | 6 | 520 | |||
GenBank mRNA | AB002386.1 | - | 5 | 488 | ||
AB004818.1 | - | 5 | 494 | |||
AK295626.1 | - | 5 | 438 | |||
AK299887.1 | - | 5 | 391 | |||
AK304835.1 | - | 5 | 389 | |||
AK310399.1 | - | 4 | 268 | |||
AK311237.1 | - | 5 | 373 | |||
AL833159.1 | - | 6 | 496 | |||
BC015882.1 | - | 6 | 510 | 3908776 | ||
BT009782.1 | - | 4 | 388 | |||
DC333017.1 | - | 6 | 533 | |||
U50315.1 | - | 5 | 494 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: U60453, AK138942, AK140694, AF483490, AF483491, and others.
Gene Ontology Categories:
