siRNA Details
Gene Symbol: EZH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321079.1 | NP_001308008.1 | 15 | 1815 | ||
NM_001321081.1 | NP_001308010.1 | 16 | 1879 | |||
NM_001321082.1 | NP_001308011.1 | 15 | 1786 | |||
NM_001991.4 | NP_001982.2 | 16 | 1906 | |||
XM_005257145.2 | XP_005257202.1 | 14 | 1661 | |||
XM_011524517.2 | XP_011522819.1 | 15 | 1754 | |||
XM_017024350.1 | XP_016879839.1 | 15 | 1733 | |||
XM_017024351.1 | XP_016879840.1 | 14 | 1640 | |||
XM_017024352.1 | XP_016879841.1 | 13 | 1491 | |||
GenBank mRNA | AB002386.1 | - | 15 | 1818 | ||
AB004818.1 | - | 15 | 1821 | |||
AK295626.1 | - | 15 | 1768 | |||
AK295853.1 | - | 14 | 1606 | |||
AK299887.1 | - | 15 | 1721 | |||
AK304835.1 | - | 15 | 1719 | |||
AK309121.1 | - | 15 | 1726 | |||
AK310399.1 | - | 14 | 1598 | |||
AK310556.1 | - | 8 | 950 | |||
AL833159.1 | - | 16 | 1826 | |||
BC015882.1 | - | 16 | 1840 | 3908776 | ||
BT009782.1 | - | 14 | 1718 | |||
U50315.1 | - | 15 | 1824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
