siRNA Details
Gene Symbol: PDPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261816.1 | NP_001248745.1 | 12 | 1939 | ||
NM_002613.4 | NP_002604.1 | 14 | 2150 | |||
NM_031268.5 | NP_112558.2 | 11 | 1769 | |||
XM_011522521.2 | XP_011520823.1 | 14 | 5838 | |||
XM_011522523.2 | XP_011520825.1 | 11 | 5462 | |||
XM_017023271.1 | XP_016878760.1 | 13 | 6218 | |||
XM_017023272.1 | XP_016878761.1 | 13 | 2774 | |||
XM_017023273.1 | XP_016878762.1 | 13 | 2483 | |||
XM_017023274.1 | XP_016878763.1 | 14 | 2621 | |||
XM_017023275.1 | XP_016878764.1 | 10 | 5843 | |||
XM_017023276.1 | XP_016878765.1 | 10 | 2087 | |||
XM_017023277.1 | XP_016878766.1 | 11 | 2185 | |||
GenBank mRNA | BC033494.1 | - | 11 | 1682 | 5176294 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
