siRNA Details
Gene Symbol: PDPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261816.1 | NP_001248745.1 | 8, 9 | 1007 | ||
NM_002613.4 | NP_002604.1 | 8, 9 | 1007 | |||
NM_031268.5 | NP_112558.2 | 5, 6 | 626 | |||
XM_011522521.2 | XP_011520823.1 | 8, 9 | 4695 | |||
XM_011522523.2 | XP_011520825.1 | 5, 6 | 4319 | |||
XM_017023271.1 | XP_016878760.1 | 8, 9 | 4700 | |||
XM_017023272.1 | XP_016878761.1 | 8, 9 | 1256 | |||
XM_017023273.1 | XP_016878762.1 | 8, 9 | 965 | |||
XM_017023274.1 | XP_016878763.1 | 9, 10 | 1103 | |||
XM_017023275.1 | XP_016878764.1 | 5, 6 | 4325 | |||
XM_017023276.1 | XP_016878765.1 | 5, 6 | 569 | |||
XM_017023277.1 | XP_016878766.1 | 6, 7 | 667 | |||
GenBank mRNA | AB209835.1 | - | 8, 9 | 974 | ||
AF017995.1 | - | 8, 9 | 938 | |||
AJ785968.1 | - | 7, 8 | 1641 | |||
AK131060.1 | - | 8, 9 | 941 | |||
AK222581.1 | - | 8, 9 | 950 | |||
BC006339.2 | - | 5, 6 | 587 | 4079551 | ||
BC012103.1 | - | 8, 9 | 967 | 4561371 | ||
BC033494.1 | - | 5, 6 | 539 | 5176294 | ||
CR536517.1 | - | 8, 9 | 858 | |||
Y15056.1 | - | 8, 9 | 937 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
