siRNA Details
Gene Symbol: PRKCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002740.5 | NP_002731.4 | 18 | 4164 | ||
XM_017006860.1 | XP_016862349.1 | 17 | 4071 | |||
GenBank mRNA | BC042405.1 | - | 6 | 2932 | ||
BM767350.1 | - | 1 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
