siRNA Details
Gene Symbol: SUV39H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282166.1 | NP_001269095.1 | 6 | 1489 | ||
NM_003173.3 | NP_003164.1 | 6 | 1242 | |||
GenBank mRNA | AF019968.1 | - | 6 | 1235 | ||
AK223071.1 | - | 6 | 1214 | |||
AK299900.1 | - | 6 | 1489 | |||
AK312547.1 | - | 6 | 1214 | |||
BC006238.2 | - | 6 | 1233 | 3945632 | ||
CR541746.1 | - | 6 | 1190 | |||
L08238.1 | - | 5 | 1330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
