siRNA Details
Gene Symbol: TTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166691.1 | NP_001160163.1 | 12 | 1495 | ||
NM_003318.4 | NP_003309.2 | 12 | 1486 | |||
XM_011536099.2 | XP_011534401.1 | 12 | 1460 | |||
XM_011536100.2 | XP_011534402.1 | 12 | 1483 | |||
XM_017011242.1 | XP_016866731.1 | 13 | 2015 | |||
GenBank mRNA | AK095599.1 | - | 12 | 1955 | ||
AK292460.1 | - | 12 | 1458 | |||
AK315696.1 | - | 12 | 1403 | |||
AX775733.1 | - | 12 | 1458 | |||
BC000633.2 | - | 12 | 1449 | 3343925 | ||
BC032858.2 | - | 12 | 1451 | 5270012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
