siRNA Details
Gene Symbol: TTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166691.1 | NP_001160163.1 | 14, 15 | 1731 | ||
NM_003318.4 | NP_003309.2 | 14, 15 | 1722 | |||
XM_011536099.2 | XP_011534401.1 | 14, 15 | 1696 | |||
XM_011536100.2 | XP_011534402.1 | 14, 15 | 1719 | |||
XM_017011242.1 | XP_016866731.1 | 15, 16 | 2251 | |||
GenBank mRNA | AK095599.1 | - | 14, 15 | 2191 | ||
AK292460.1 | - | 14, 15 | 1694 | |||
AK315696.1 | - | 14, 15 | 1639 | |||
AX775733.1 | - | 14, 15 | 1694 | |||
BC000633.2 | - | 14, 15 | 1685 | 3343925 | ||
BC032858.2 | - | 14, 15 | 1687 | 5270012 | ||
X70500.1 | - | 1, 2 | 144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
