siRNA Details
Gene Symbol: UBE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282161.1 | NP_001269090.1 | 6 | 1558 | ||
NM_003336.3 | NP_003327.2 | 6 | 1565 | |||
NM_181762.2 | NP_861427.1 | 5 | 1475 | |||
GenBank mRNA | AK223045.1 | - | 6 | 1467 | ||
AK223064.1 | - | 6 | 1464 | |||
BC042021.1 | - | 3 | 2342 | |||
M74524.1 | - | 6 | 1440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
