siRNA Details
Gene Symbol: UBE2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003339.2 | NP_003330.1 | 7 | 2431 | ||
NM_181838.1 | NP_862821.1 | 8 | 2595 | |||
GenBank mRNA | AF317220.1 | - | 5 | 1727 | ||
AK001428.1 | - | 7 | 2258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
