siRNA Details
Gene Symbol: UBE2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003339.2 | NP_003330.1 | 5 | 920 | ||
NM_181838.1 | NP_862821.1 | 6 | 1084 | |||
XM_017009820.1 | XP_016865309.1 | 8 | 1291 | |||
GenBank mRNA | AF317220.1 | - | 3 | 228 | ||
AK001311.1 | - | 5 | 693 | |||
AK001428.1 | - | 5 | 747 | |||
AK310273.1 | - | 5 | 649 | |||
AY651263.1 | - | 5 | 676 | |||
BC019052.2 | - | 5 | 704 | |||
BC033349.1 | - | 5 | 920 | 4827210 | ||
U39317.1 | - | 5 | 316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
