siRNA Details
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 8 | 1241 | ||
NM_003340.6 | NP_003331.1 | 8 | 1548 | |||
NM_181886.3 | NP_871615.1 | 8 | 1756 | |||
NM_181887.2 | NP_871616.1 | 8 | 1526 | |||
NM_181888.3 | NP_871617.1 | 8 | 1405 | |||
NM_181889.2 | NP_871618.1 | 8 | 1428 | |||
NM_181890.2 | NP_871619.1 | 8 | 1370 | |||
NM_181891.2 | NP_871620.1 | 8 | 1713 | |||
NM_181892.3 | NP_871621.1 | 8 | 1788 | |||
NM_181893.2 | NP_871622.1 | 7 | 1218 | |||
XM_017008584.1 | XP_016864073.1 | 7 | 1580 | |||
GenBank mRNA | AK095822.1 | - | 8 | 1703 | ||
AK127304.1 | - | 6 | 4161 | |||
AL050186.1 | - | 1 | 91 | |||
BC003395.1 | - | 8 | 1228 | 3447673 | ||
BC037894.2 | - | 8 | 1526 | 5275356 | ||
BC066917.1 | - | 8 | 1259 | 4792462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
