siRNA Details
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 8 | 1713 | ||
NM_003340.6 | NP_003331.1 | 8 | 2020 | |||
NM_181886.3 | NP_871615.1 | 8 | 2228 | |||
NM_181887.2 | NP_871616.1 | 8 | 1998 | |||
NM_181888.3 | NP_871617.1 | 8 | 1877 | |||
NM_181889.2 | NP_871618.1 | 8 | 1900 | |||
NM_181890.2 | NP_871619.1 | 8 | 1842 | |||
NM_181891.2 | NP_871620.1 | 8 | 2185 | |||
NM_181892.3 | NP_871621.1 | 8 | 2260 | |||
NM_181893.2 | NP_871622.1 | 7 | 1690 | |||
XM_017008584.1 | XP_016864073.1 | 7 | 2052 | |||
GenBank mRNA | AK095822.1 | - | 8 | 2175 | ||
AK127304.1 | - | 6 | 4633 | |||
AL050186.1 | - | 1 | 563 | |||
BC037894.2 | - | 8 | 1998 | 5275356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
