siRNA Details
Gene Symbol: UBE2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202476.1 | NP_001189405.1 | 5 | 1111 | ||
NM_003341.4 | NP_003332.1 | 6 | 1406 | |||
NM_182666.2 | NP_872607.1 | 5 | 1355 | |||
GenBank mRNA | BC009139.1 | - | 6 | 1326 | 3853408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
