siRNA Details
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003345.4 | NP_003336.1 | 5, 6 | 499 | ||
NM_194259.2 | NP_919235.1 | 6, 7 | 756 | |||
NM_194260.2 | NP_919236.1 | 5, 6 | 619 | |||
NM_194261.2 | NP_919237.1 | 5, 6 | 471 | |||
XM_005255540.2 | XP_005255597.1 | 5, 6 | 490 | |||
XM_005255541.4 | XP_005255598.1 | 5, 6 | 896 | |||
XM_011522644.2 | XP_011520946.1 | 6, 7 | 736 | |||
XM_011522645.1 | XP_011520947.1 | 5, 6 | 601 | |||
XM_017023640.1 | XP_016879129.1 | 5, 6 | 896 | |||
GenBank mRNA | AB208988.1 | - | 5, 6 | 580 | ||
AJ002385.1 | - | 5, 6 | 419 | |||
AK024172.1 | - | 1 | 1191 | |||
AK291118.1 | - | 5, 6 | 490 | |||
BC000427.2 | - | 5, 6 | 403 | 2819764 | ||
BC004429.1 | - | 5, 6 | 403 | 2819764 | ||
BC051289.1 | - | 5, 6 | 433 | |||
BI523501.1 | - | 5, 6 | 609 | |||
BT006932.1 | - | 4, 5 | 341 | |||
BT007010.1 | - | 4, 5 | 341 | |||
BU146357.1 | - | 1, 2 | 72 | |||
BU543455.1 | - | 5, 6 | 491 | |||
CD249028.1 | - | 5, 6 | 440 | |||
CX788607.1 | - | 5, 6 | 575 | |||
D45050.1 | - | 5, 6 | 458 | |||
U29092.1 | - | 4, 5 | 365 | |||
U31882.1 | - | 5, 6 | 434 | |||
U31933.1 | - | 5, 6 | 459 | |||
U38785.1 | - | 5, 6 | 433 | |||
U45328.1 | - | 5, 6 | 464 | |||
U66818.1 | - | 5, 6 | 428 | |||
U66867.1 | - | 5, 6 | 1147 | |||
X96427.1 | - | 5, 6 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
