siRNA Details
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003345.4 | NP_003336.1 | 7 | 625 | ||
NM_194259.2 | NP_919235.1 | 8 | 882 | |||
NM_194260.2 | NP_919236.1 | 7 | 745 | |||
NM_194261.2 | NP_919237.1 | 7 | 597 | |||
XM_017023640.1 | XP_016879129.1 | 7 | 1022 | |||
GenBank mRNA | AB208988.1 | - | 7 | 706 | ||
AJ002385.1 | - | 7 | 545 | |||
AK291118.1 | - | 7 | 616 | |||
BC000427.2 | - | 7 | 529 | 2819764 | ||
BC004429.1 | - | 7 | 529 | 2819764 | ||
BC051289.1 | - | 7 | 559 | |||
BQ230918.1 | - | 2 | 115 | |||
BU146357.1 | - | 3 | 198 | |||
BU543455.1 | - | 7 | 617 | |||
CD249028.1 | - | 7 | 566 | |||
CX788607.1 | - | 7 | 701 | |||
D45050.1 | - | 7 | 584 | |||
U29092.1 | - | 6 | 491 | |||
U31882.1 | - | 7 | 560 | |||
U31933.1 | - | 7 | 585 | |||
U38785.1 | - | 7 | 559 | |||
U45328.1 | - | 7 | 590 | |||
U66818.1 | - | 7 | 554 | |||
U66867.1 | - | 7 | 1273 | |||
X96427.1 | - | 7 | 590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
