siRNA Details
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003345.4 | NP_003336.1 | 4 | 362 | ||
NM_194259.2 | NP_919235.1 | 5 | 619 | |||
NM_194260.2 | NP_919236.1 | 4 | 482 | |||
NM_194261.2 | NP_919237.1 | 4 | 334 | |||
XM_005255540.2 | XP_005255597.1 | 4 | 353 | |||
XM_005255541.4 | XP_005255598.1 | 4 | 759 | |||
XM_011522644.2 | XP_011520946.1 | 5 | 599 | |||
XM_011522645.1 | XP_011520947.1 | 4 | 464 | |||
XM_017023640.1 | XP_016879129.1 | 4 | 759 | |||
GenBank mRNA | AB208988.1 | - | 4 | 443 | ||
AJ002385.1 | - | 4 | 282 | |||
AK093616.1 | - | 4 | 293 | |||
AK291118.1 | - | 4 | 353 | |||
AL833737.1 | - | 1 | 1159 | |||
BC000427.2 | - | 4 | 266 | 2819764 | ||
BC004429.1 | - | 4 | 266 | 2819764 | ||
BC051289.1 | - | 4 | 296 | |||
BE262845.1 | - | 5 | 601 | |||
BI523501.1 | - | 4 | 472 | |||
BT006932.1 | - | 3 | 204 | |||
BT007010.1 | - | 3 | 204 | |||
BU543455.1 | - | 4 | 354 | |||
CD249028.1 | - | 4 | 303 | |||
CX788607.1 | - | 4 | 438 | |||
D45050.1 | - | 4 | 321 | |||
U29092.1 | - | 3 | 228 | |||
U31882.1 | - | 4 | 297 | |||
U31933.1 | - | 4 | 322 | |||
U38785.1 | - | 4 | 296 | |||
U45328.1 | - | 4 | 327 | |||
U66818.1 | - | 4 | 291 | |||
U66867.1 | - | 4 | 1010 | |||
X96427.1 | - | 4 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
