siRNA Details
Gene Symbol: UBE2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256355.1 | NP_001243284.1 | 4 | 723 | ||
NM_001256356.1 | NP_001243285.1 | 3 | 648 | |||
NM_003347.3 | NP_003338.1 | 4 | 744 | |||
NR_028436.2 | 5 | 853 | ||||
NR_046082.1 | 4 | 1167 | ||||
GenBank mRNA | AJ000519.1 | - | 4 | 561 | ||
AK125396.1 | - | 4 | 1167 | |||
AK291083.1 | - | 4 | 556 | |||
AK293179.1 | - | 3 | 458 | |||
AK299985.1 | - | 4 | 723 | |||
BC053368.1 | - | 4 | 597 | 4214359 | ||
BU567707.1 | - | 5 | 673 | |||
S81003.1 | - | 4 | 561 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009456, BC093503, AK007265, AK167847, AK165620, and others.
Gene Ontology Categories:
