siRNA Details
Gene Symbol: UQCRQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014402.4 | NP_055217.2 | 3 | 385 | ||
GenBank mRNA | AK297061.1 | - | 2 | 822 | ||
AW327741.1 | - | 2 | 276 | |||
BC001390.2 | - | 2 | 338 | 3050489 | ||
BC064949.1 | - | 3 | 359 | |||
BC090048.1 | - | 3 | 362 | 30393278 | ||
BU953465.1 | - | 3 | 388 | |||
CR743513.1 | - | 1 | 92 |
Gene Symbol: ZNF740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC053557.1 | - | 2067 | 6162269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
