siRNA Details
Gene Symbol: ST6GALNAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175039.3 | NP_778204.1 | 6 | 1669 | ||
NM_175040.3 | NP_778205.1 | 5 | 1582 | |||
XM_011518527.2 | XP_011516829.1 | 7 | 1761 | |||
XM_011518529.2 | XP_011516831.1 | 7 | 1792 | |||
GenBank mRNA | AK292049.1 | - | 6 | 1424 | ||
BC036705.2 | - | 6 | 1572 | 4309208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
