siRNA Details
Gene Symbol: UBE2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256355.1 | NP_001243284.1 | 4 | 2283 | ||
NM_001256356.1 | NP_001243285.1 | 3 | 2208 | |||
NM_003347.3 | NP_003338.1 | 4 | 2304 | |||
NR_028436.2 | 5 | 2413 | ||||
NR_046082.1 | 4 | 2727 | ||||
GenBank mRNA | AJ000519.1 | - | 4 | 2121 | ||
AK125396.1 | - | 4 | 2727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
