siRNA Details
Gene Symbol: UBE2N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003348.3 | NP_003339.1 | 4 | 1273 | ||
GenBank mRNA | AK098233.1 | - | 1 | 1708 | ||
BC000396.2 | - | 4 | 999 | 2822013 | ||
BC003365.1 | - | 4 | 934 | 2900313 | ||
BC108704.1 | - | 4 | 968 | 6503770 | ||
D83004.1 | - | 4 | 970 | |||
FJ224354.1 | - | 4 | 970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
