siRNA Details
Gene Symbol: UBE2N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003348.3 | NP_003339.1 | 4 | 1619 | ||
GenBank mRNA | AK098233.1 | - | 1 | 2055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
