siRNA Details
Gene Symbol: HDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003883.3 | NP_003874.2 | 7 | 640 | ||
XM_011537697.2 | XP_011535999.1 | 6 | 492 | |||
GenBank mRNA | AF005482.1 | - | 5 | 656 | ||
AF039703.1 | - | 7 | 596 | |||
AY429537.1 | - | 3 | 208 | |||
AY429538.1 | - | 5 | 323 | |||
BC000614.1 | - | 7 | 629 | 3345755 | ||
BM461585.1 | - | 7 | 630 | |||
U66914.1 | - | 7 | 585 | |||
U75696.1 | - | 6 | 623 | |||
U75697.1 | - | 7 | 629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
