siRNA Details
Gene Symbol: HERC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003922.3 | NP_003913.3 | 5 | 1512 | ||
XM_017022699.1 | XP_016878188.1 | 5 | 1525 | |||
XM_017022700.1 | XP_016878189.1 | 5 | 1525 | |||
XM_017022701.1 | XP_016878190.1 | 5 | 1525 | |||
XM_017022702.1 | XP_016878191.1 | 5 | 1525 | |||
XM_017022703.1 | XP_016878192.1 | 5 | 1525 | |||
XM_017022704.1 | XP_016878193.1 | 5 | 1525 | |||
XM_017022705.1 | XP_016878194.1 | 5 | 1525 | |||
XM_017022706.1 | XP_016878195.1 | 5 | 1528 | |||
GenBank mRNA | AK310519.1 | - | 5 | 1481 | ||
U50078.1 | - | 5 | 1460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
