siRNA Details
Gene Symbol: UBE2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003969.3 | NP_003960.1 | 2 | 786 | ||
GenBank mRNA | AB012191.1 | - | 2 | 191 | ||
AF075599.1 | - | 2 | 194 | |||
BC058924.1 | - | 2 | 300 | 6162220 | ||
BT006754.1 | - | 2 | 191 |
Gene Symbol: UBE2MP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW376368.1 | - | 1 | 142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
