siRNA Details
Gene Symbol: DDX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320595.1 | NP_001307524.1 | 14 | 2239 | ||
NM_001320596.1 | NP_001307525.1 | 14 | 2128 | |||
NM_001320597.1 | NP_001307526.1 | 13 | 1979 | |||
NM_004396.4 | NP_004387.1 | 13 | 2214 | |||
GenBank mRNA | AB209919.1 | - | 12 | 3201 | ||
AB451257.1 | - | 13 | 1812 | |||
AB451382.1 | - | 13 | 1812 | |||
AK055995.1 | - | 2 | 1312 | |||
AK297192.1 | - | 12 | 1744 | |||
AK297753.1 | - | 14 | 1949 | |||
AK297905.1 | - | 12 | 1772 | |||
AK310318.1 | - | 13 | 1858 | |||
BC016027.1 | - | 13 | 1953 | 3528578 | ||
BT006943.1 | - | 13 | 1812 | |||
BU160611.1 | - | 72 | ||||
BX571764.1 | - | 12 | 3219 | |||
X15729.1 | - | 13 | 1979 | |||
X52104.1 | - | 13 | 1987 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
