siRNA Details
Gene Symbol: DDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257191.2 | NP_001244120.1 | 14 | 2445 | ||
NM_004397.5 | NP_004388.2 | 14 | 2517 | |||
XM_005271417.3 | XP_005271474.1 | 14 | 2462 | |||
XM_011542644.1 | XP_011540946.1 | 13 | 2388 | |||
XM_011542645.1 | XP_011540947.1 | 13 | 2370 | |||
XM_017017251.1 | XP_016872740.1 | 13 | 2333 | |||
GenBank mRNA | AK093657.1 | - | 6 | 1687 | ||
BC039826.1 | - | 7 | 1298 | |||
BC065007.1 | - | 13 | 2375 | 6163439 | ||
D17532.1 | - | 14 | 2386 | |||
Z11685.1 | - | 14 | 2417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
