siRNA Details
Gene Symbol: HERC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004667.5 | NP_004658.3 | 3, 4 | 288 | ||
XM_005268276.4 | XP_005268333.1 | 2, 3 | 418 | |||
XM_006720726.3 | XP_006720789.1 | 3, 4 | 309 | |||
XM_006720727.3 | XP_006720790.1 | 3, 4 | 309 | |||
XM_011522131.2 | XP_011520433.1 | 1, 2 | 74 | |||
XM_011522133.2 | XP_011520435.1 | 3, 4 | 309 | |||
XM_017022695.1 | XP_016878184.1 | 2, 3 | 235 | |||
XM_017022696.1 | XP_016878185.1 | 2, 3 | 178 | |||
GenBank mRNA | AF071172.3 | - | 3, 4 | 240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
