siRNA Details
Gene Symbol: HERC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004667.5 | NP_004658.3 | 48 | 7777 | ||
XM_005268276.4 | XP_005268333.1 | 47 | 7907 | |||
XM_006720726.3 | XP_006720789.1 | 48 | 7783 | |||
XM_006720727.3 | XP_006720790.1 | 46 | 7540 | |||
XM_011522131.2 | XP_011520433.1 | 46 | 7564 | |||
XM_011522133.2 | XP_011520435.1 | 29 | 4543 | |||
XM_017022695.1 | XP_016878184.1 | 47 | 7724 | |||
XM_017022696.1 | XP_016878185.1 | 47 | 7667 | |||
XM_017022697.1 | XP_016878186.1 | 7 | 1106 | |||
XM_017022698.1 | XP_016878187.1 | 7 | 1017 | |||
GenBank mRNA | AF071172.3 | - | 48 | 7729 | ||
AK122977.1 | - | 3 | 1778 | |||
BC148318.2 | - | 2 | 71 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
