siRNA Details
Gene Symbol: HDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004964.2 | NP_004955.2 | 9 | 914 | ||
XM_011541309.2 | XP_011539611.1 | 9 | 1299 | |||
GenBank mRNA | AB451297.1 | - | 9 | 851 | ||
AB451430.1 | - | 9 | 851 | |||
AK299241.1 | - | 9 | 848 | |||
AK313061.1 | - | 9 | 916 | |||
AY627042.1 | - | 9 | 858 | |||
BC000301.2 | - | 9 | 885 | 2820260 | ||
BX648055.1 | - | 2 | 1221 | |||
CR541717.1 | - | 9 | 851 | |||
D50405.1 | - | 9 | 914 | |||
U50079.1 | - | 9 | 961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
