siRNA Details
Gene Symbol: PLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005030.5 | NP_005021.2 | 9 | 1755 | ||
GenBank mRNA | AB209179.1 | - | 8 | 3614 | ||
AK303263.1 | - | 10 | 1371 | |||
AK313227.1 | - | 9 | 1679 | |||
BC002369.2 | - | 9 | 1647 | 2822226 | ||
BC003002.1 | - | 9 | 1625 | 2823623 | ||
BC014846.1 | - | 9 | 1600 | 3854860 | ||
L19559.1 | - | 9 | 1657 | |||
U01038.1 | - | 9 | 1657 | |||
X73458.1 | - | 9 | 1671 | |||
X75932.1 | - | 9 | 1657 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
