siRNA Details
Gene Symbol: USP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272075.1 | NP_001259004.1 | 7, 8 | 1662 | ||
NM_005153.2 | NP_005144.2 | 6, 7 | 1541 | |||
NR_073577.1 | 4, 5 | 439 | ||||
XM_006721332.1 | XP_006721395.1 | 7, 8 | 1548 | |||
XM_011523440.1 | XP_011521742.1 | 8, 9 | 1669 | |||
XM_011523441.1 | XP_011521743.1 | 9, 10 | 1993 | |||
XM_011523443.1 | XP_011521745.1 | 8, 9 | 1740 | |||
XM_017023863.1 | XP_016879352.1 | 7, 8 | 1687 | |||
XM_017023864.1 | XP_016879353.1 | 10, 11 | 1963 | |||
XM_017023865.1 | XP_016879354.1 | 9, 10 | 1832 | |||
XM_017023866.1 | XP_016879355.1 | 7, 8 | 2364 | |||
XM_017023867.1 | XP_016879356.1 | 9, 10 | 1955 | |||
XM_017023868.1 | XP_016879357.1 | 8, 9 | 1789 | |||
XM_017023869.1 | XP_016879358.1 | 8, 9 | 1826 | |||
GenBank mRNA | AK293304.1 | - | 4, 5 | 384 | ||
AK299618.1 | - | 7, 8 | 1607 | |||
AK315570.1 | - | 6, 7 | 1495 | |||
AL162049.1 | - | 6, 7 | 1480 | |||
BC000263.1 | - | 6, 7 | 1512 | 3501606 | ||
BX537402.1 | - | 6, 7 | 1479 | |||
D80012.1 | - | 6, 7 | 1445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
