siRNA Details
Gene Symbol: USP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272075.1 | NP_001259004.1 | 10 | 1882 | ||
NM_005153.2 | NP_005144.2 | 9 | 1761 | |||
NR_073577.1 | 7 | 659 | ||||
NR_073578.1 | 6 | 1498 | ||||
XM_006721332.1 | XP_006721395.1 | 10 | 1768 | |||
XM_011523440.1 | XP_011521742.1 | 11 | 1889 | |||
XM_011523441.1 | XP_011521743.1 | 12 | 2213 | |||
XM_011523443.1 | XP_011521745.1 | 11 | 1960 | |||
XM_017023863.1 | XP_016879352.1 | 10 | 1907 | |||
XM_017023864.1 | XP_016879353.1 | 13 | 2183 | |||
XM_017023865.1 | XP_016879354.1 | 12 | 2052 | |||
XM_017023866.1 | XP_016879355.1 | 10 | 2584 | |||
XM_017023867.1 | XP_016879356.1 | 12 | 2175 | |||
XM_017023868.1 | XP_016879357.1 | 11 | 2009 | |||
XM_017023869.1 | XP_016879358.1 | 11 | 2046 | |||
GenBank mRNA | AK293304.1 | - | 7 | 604 | ||
AK299618.1 | - | 10 | 1827 | |||
AK315570.1 | - | 9 | 1715 | |||
AL162049.1 | - | 9 | 1700 | |||
BC000263.1 | - | 9 | 1732 | 3501606 | ||
BX537402.1 | - | 9 | 1699 | |||
CR749515.1 | - | 6 | 1453 | |||
D80012.1 | - | 9 | 1665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
