siRNA Details
Gene Symbol: USP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272075.1 | NP_001259004.1 | 7 | 1632 | ||
NM_005153.2 | NP_005144.2 | 6 | 1511 | |||
NR_073577.1 | 4 | 409 | ||||
XM_006721332.1 | XP_006721395.1 | 7 | 1518 | |||
XM_011523440.1 | XP_011521742.1 | 8 | 1639 | |||
XM_011523441.1 | XP_011521743.1 | 9 | 1963 | |||
XM_011523443.1 | XP_011521745.1 | 8 | 1710 | |||
XM_017023863.1 | XP_016879352.1 | 7 | 1657 | |||
XM_017023864.1 | XP_016879353.1 | 10 | 1933 | |||
XM_017023865.1 | XP_016879354.1 | 9 | 1802 | |||
XM_017023866.1 | XP_016879355.1 | 7 | 2334 | |||
XM_017023867.1 | XP_016879356.1 | 9 | 1925 | |||
XM_017023868.1 | XP_016879357.1 | 8 | 1759 | |||
XM_017023869.1 | XP_016879358.1 | 8 | 1796 | |||
GenBank mRNA | AK293304.1 | - | 4 | 354 | ||
AK299618.1 | - | 7 | 1577 | |||
AK315570.1 | - | 6 | 1465 | |||
AL162049.1 | - | 6 | 1450 | |||
BC000263.1 | - | 6 | 1482 | 3501606 | ||
BX537402.1 | - | 6 | 1449 | |||
D80012.1 | - | 6 | 1415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
