siRNA Details
Gene Symbol: HDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015053.1 | NP_001015053.1 | 12 | 1873 | ||
NM_005474.4 | NP_005465.2 | 12 | 1870 | |||
XM_005256905.1 | XP_005256962.1 | 12 | 1868 | |||
XM_005256906.4 | XP_005256963.1 | 12 | 1829 | |||
XM_011524149.1 | XP_011522451.1 | 12 | 1844 | |||
XM_011524150.1 | XP_011522452.1 | 11 | 1620 | |||
XM_017023988.1 | XP_016879477.1 | 14 | 1982 | |||
XM_017023989.1 | XP_016879478.1 | 14 | 1948 | |||
XM_017023990.1 | XP_016879479.1 | 14 | 1973 | |||
XM_017023991.1 | XP_016879480.1 | 14 | 1979 | |||
XM_017023992.1 | XP_016879481.1 | 14 | 1968 | |||
XM_017023993.1 | XP_016879482.1 | 14 | 1955 | |||
XM_017023994.1 | XP_016879483.1 | 10 | 1936 | |||
XM_017023995.1 | XP_016879484.1 | 12 | 1838 | |||
XM_017023996.1 | XP_016879485.1 | 13 | 2045 | |||
GenBank mRNA | AB011172.2 | - | 12 | 1842 | ||
AB177853.1 | - | 2 | 322 | |||
AF039691.1 | - | 7 | 975 | |||
AF132608.1 | - | 11 | 1538 | |||
AK075095.1 | - | 3 | 519 | |||
AK294760.1 | - | 12 | 1838 | |||
BC038423.1 | - | 1 | 152 | |||
BC051824.1 | - | 12 | 1749 | 6043491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
