siRNA Details
Gene Symbol: HDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015053.1 | NP_001015053.1 | 18, 19 | 2800 | ||
NM_005474.4 | NP_005465.2 | 18, 19 | 2797 | |||
XM_005256905.1 | XP_005256962.1 | 18, 19 | 2795 | |||
XM_005256906.4 | XP_005256963.1 | 18, 19 | 2756 | |||
XM_011524149.1 | XP_011522451.1 | 18, 19 | 2771 | |||
XM_011524150.1 | XP_011522452.1 | 17, 18 | 2547 | |||
XM_017023988.1 | XP_016879477.1 | 20, 21 | 2909 | |||
XM_017023989.1 | XP_016879478.1 | 20, 21 | 2875 | |||
XM_017023990.1 | XP_016879479.1 | 20, 21 | 2900 | |||
XM_017023991.1 | XP_016879480.1 | 20, 21 | 2906 | |||
XM_017023992.1 | XP_016879481.1 | 20, 21 | 2895 | |||
XM_017023993.1 | XP_016879482.1 | 20, 21 | 2882 | |||
XM_017023994.1 | XP_016879483.1 | 16, 17 | 2863 | |||
XM_017023995.1 | XP_016879484.1 | 18, 19 | 2765 | |||
XM_017023996.1 | XP_016879485.1 | 19, 20 | 2972 | |||
GenBank mRNA | AB011172.2 | - | 16, 17 | 2514 | ||
AB177853.1 | - | 8, 9 | 1249 | |||
AF039691.1 | - | 13, 14 | 1902 | |||
AF132608.1 | - | 17, 18 | 2465 | |||
AK027734.1 | - | 4, 5 | 615 | |||
AK075095.1 | - | 9, 10 | 1445 | |||
AK294760.1 | - | 18, 19 | 2765 | |||
BC038423.1 | - | 7, 8 | 1079 | |||
BC051824.1 | - | 18, 19 | 2676 | 6043491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
