siRNA Details
Gene Symbol: HDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015053.1 | NP_001015053.1 | 8 | 1136 | ||
NM_005474.4 | NP_005465.2 | 8 | 1133 | |||
XM_005256905.1 | XP_005256962.1 | 8 | 1131 | |||
XM_005256906.4 | XP_005256963.1 | 8 | 1092 | |||
XM_011524149.1 | XP_011522451.1 | 8 | 1107 | |||
XM_011524150.1 | XP_011522452.1 | 7 | 883 | |||
XM_017023988.1 | XP_016879477.1 | 10 | 1245 | |||
XM_017023989.1 | XP_016879478.1 | 10 | 1211 | |||
XM_017023990.1 | XP_016879479.1 | 10 | 1236 | |||
XM_017023991.1 | XP_016879480.1 | 10 | 1242 | |||
XM_017023992.1 | XP_016879481.1 | 10 | 1246 | |||
XM_017023993.1 | XP_016879482.1 | 10 | 1218 | |||
XM_017023994.1 | XP_016879483.1 | 6 | 1199 | |||
XM_017023995.1 | XP_016879484.1 | 8 | 1101 | |||
XM_017023996.1 | XP_016879485.1 | 9 | 1308 | |||
GenBank mRNA | AB011172.2 | - | 8 | 1105 | ||
AF039691.1 | - | 3 | 238 | |||
AF132608.1 | - | 7 | 801 | |||
AK294760.1 | - | 8 | 1101 | |||
BC013140.1 | - | 5 | 583 | |||
BC051824.1 | - | 8 | 1012 | 6043491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
