siRNA Details
Gene Symbol: G3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005754.2 | NP_005745.1 | 12 | 2024 | ||
NM_198395.1 | NP_938405.1 | 12 | 1993 | |||
XM_006714749.3 | XP_006714812.1 | 12 | 2013 | |||
XM_006714750.3 | XP_006714813.1 | 12 | 1952 | |||
XM_017008922.1 | XP_016864411.1 | 12 | 2299 | |||
XM_017008923.1 | XP_016864412.1 | 12 | 2268 | |||
GenBank mRNA | AK130003.1 | - | 9 | 1508 | ||
AK222606.1 | - | 12 | 1987 | |||
BC006997.1 | - | 12 | 1988 | 3950738 | ||
BC108278.1 | - | 12 | 1931 | 6051310 | ||
BX647869.1 | - | 11 | 1895 | |||
BX648012.1 | - | 12 | 2001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
