siRNA Details
Gene Symbol: PSMD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005805.5 | NP_005796.1 | 11 | 1282 | ||
GenBank mRNA | AB209220.1 | - | 4 | 4458 | ||
AK055128.1 | - | 11 | 1519 | |||
AK091095.1 | - | 1 | 1353 | |||
BC009524.1 | - | 2 | 172 | |||
BC066336.1 | - | 11 | 1128 | 5296432 | ||
U86782.1 | - | 11 | 1014 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
