siRNA Details
Gene Symbol: PSMD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005805.5 | NP_005796.1 | 12 | 1330 | ||
GenBank mRNA | AB209220.1 | - | 5 | 4506 | ||
AK055128.1 | - | 12 | 1567 | |||
AK091095.1 | - | 2 | 1401 | |||
BC009524.1 | - | 3 | 220 | |||
BC066336.1 | - | 12 | 1176 | 5296432 | ||
U86782.1 | - | 12 | 1062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
