siRNA Details
Gene Symbol: MCM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278595.1 | NP_001265524.1 | 10 | 2235 | ||
NM_005916.4 | NP_005907.3 | 11 | 2160 | |||
NM_182776.2 | NP_877577.1 | 10 | 2126 | |||
XM_005250348.3 | XP_005250405.1 | 11 | 1814 | |||
XM_017012217.1 | XP_016867706.1 | 11 | 1691 | |||
GenBank mRNA | AF279900.1 | - | 2 | 204 | ||
AK055379.1 | - | 10 | 2103 | |||
AK096959.1 | - | 12 | 1524 | |||
AK226175.1 | - | 10 | 1665 | |||
AK293172.1 | - | 11 | 1562 | |||
AK314575.1 | - | 11 | 1627 | |||
AY007130.1 | - | 3 | 457 | |||
BC013375.2 | - | 11 | 1596 | 3946342 | ||
D55716.1 | - | 11 | 1630 | |||
D86748.1 | - | 11 | 1600 | |||
KU178172.1 | - | 11 | 1514 | |||
KU178173.1 | - | 10 | 1589 | |||
X74796.1 | - | 8 | 1210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
